The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204077.1 | NP_001191006.1 | 17 - 18 | 2903 | 76 |
NM_004788.3 | NP_004779.2 | 17 - 18 | 2924 | 76 | |
GenBank mRNA | AK296369.1 | - | 17 - 18 | 2850 | 76 |
AK302328.1 | - | 8 - 9 | 1649 | 76 | |
BC111417.1 | - | 17 - 18 | 2877 | 76 | |
BC112367.1 | - | 17 - 18 | 2880 | 76 | |
D50916.1 | - | 17 - 18 | 2861 | 76 |