Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAM
Entrez Gene ID
5066
Gene Name
peptidylglycine alpha-amidating monooxygenase
Gene Aliases
PAL, PHM
Chromosome Location
Chr.5: 102754783 - 103031105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000919.3 NP_000910.2 13 - 14 1532 69
NM_001177306.1 NP_001170777.1 13 - 14 1532 69
NM_001319943.1 NP_001306872.1 14 - 15 2098 69
NM_138766.2 NP_620121.1 13 - 14 1532 69
NM_138822.2 NP_620177.1 13 - 14 1532 69
NR_033440.1 12 - 13 1474 69
XM_011543419.2 XP_011541721.1 14 - 15 1652 69
XM_017009495.1 XP_016864984.1 13 - 14 1528 69
XM_017009496.1 XP_016864985.1 13 - 14 1528 69
XM_017009497.1 XP_016864986.1 14 - 15 1652 69
XM_017009498.1 XP_016864987.1 13 - 14 1528 69
XM_017009499.1 XP_016864988.1 13 - 14 1528 69
XM_017009500.1 XP_016864989.1 13 - 14 1528 69
XM_017009501.1 XP_016864990.1 13 - 14 1528 69
XM_017009502.1 XP_016864991.1 13 - 14 1528 69
XM_017009503.1 XP_016864992.1 13 - 14 1528 69
XM_017009504.1 XP_016864993.1 13 - 14 1629 69
XM_017009505.1 XP_016864994.1 14 - 15 1652 69
XM_017009506.1 XP_016864995.1 13 - 14 1528 69
XM_017009511.1 XP_016865000.1 13 - 14 1528 69
XM_017009512.1 XP_016865001.1 13 - 14 1528 69
XM_017009513.1 XP_016865002.1 13 - 14 1528 69
GenBank mRNA AB095007.1 - 13 - 14 1159 69
AF010472.1 - 12 - 13 1075 69
AK290158.1 - 13 - 14 1270 69
AK296521.1 - 12 - 13 1240 69
M37721.1 - 13 - 14 1347 69
S75037.1 - 13 - 14 1239 69
S75038.1 - 13 - 14 1239 69

View Full Product Details