The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015913.3 | NP_056997.1 | 2 - 3 | 1230 | 82 |
GenBank mRNA | AF131758.1 | - | 2 - 3 | 265 | 82 |
AF543416.1 | - | 2 - 3 | 434 | 82 | |
AK056169.1 | - | 2 - 3 | 1230 | 82 | |
AK075409.1 | - | 2 - 3 | 228 | 82 | |
AY358982.1 | - | 2 - 3 | 261 | 82 | |
BC001493.2 | - | 2 - 3 | 231 | 82 | |
BC008913.2 | - | 2 - 3 | 260 | 82 | |
BC008953.2 | - | 2 - 3 | 209 | 82 |