The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015913.3 | NP_056997.1 | 3 - 4 | 1289 | 141 |
GenBank mRNA | AF131758.1 | - | 3 - 4 | 324 | 141 |
AF543416.1 | - | 3 - 4 | 493 | 141 | |
AK056169.1 | - | 3 - 4 | 141 | ||
AK075409.1 | - | 3 - 4 | 287 | 141 | |
AY358982.1 | - | 3 - 4 | 320 | 141 | |
BC001493.2 | - | 3 - 4 | 290 | 141 | |
BC008913.2 | - | 3 - 4 | 319 | 141 | |
BC008953.2 | - | 3 - 4 | 268 | 141 |