Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGC
Entrez Gene ID
5225
Gene Name
progastricsin
Gene Aliases
PEPC, PGII
Chromosome Location
Chr.6: 41736711 - 41747401 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166424.1 NP_001159896.1 3 - 4 392 90
NM_002630.3 NP_002621.1 3 - 4 392 90
GenBank mRNA AK223274.1 - 3 - 4 388 90
AK301298.1 - 3 - 4 388 90
AK301325.1 - 3 - 4 388 90
BC073740.1 - 3 - 4 374 90
J04443.1 - 3 - 4 332 90
U75272.1 - 3 - 4 375 90

View Full Product Details