The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166424.1 | NP_001159896.1 | 4 - 5 | 513 | 70 |
NM_002630.3 | NP_002621.1 | 4 - 5 | 513 | 70 | |
GenBank mRNA | AK223274.1 | - | 4 - 5 | 509 | 70 |
AK301298.1 | - | 4 - 5 | 509 | 70 | |
BC073740.1 | - | 4 - 5 | 495 | 70 | |
J04443.1 | - | 4 - 5 | 453 | 70 | |
U75272.1 | - | 4 - 5 | 496 | 70 |