Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPIFB2
Entrez Gene ID
80341
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2
Chromosome Location
Chr.20: 33007578 - 33023709 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025227.2 NP_079503.1 10 - 11 1127 77
GenBank mRNA AF465765.1 - 9 - 10 910 77
AK027068.1 - 10 - 11 1105 77
AK172819.1 - 10 - 11 1106 77
AK304064.1 - 10 - 11 957 77
AY358723.1 - 10 - 11 1035 77
BC034415.1 - 10 - 11 981 77

View Full Product Details