Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDZD3
Entrez Gene ID
79849
Gene Name
PDZ domain containing 3
Gene Aliases
IKEPP, NHERF4, PDZK2
Chromosome Location
Chr.11: 119185457 - 119190223 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168468.1 NP_001161940.1 1 - 2 54 64
NM_024791.3 NP_079067.3 1 - 2 54 64
NR_033122.1 1 - 2 54 64
XM_011543000.2 XP_011541302.1 1 - 2 36 64
GenBank mRNA AK026409.1 - 1 - 2 36 64
AK091966.1 - 1 - 2 36 64
AK298333.1 - 1 - 2 36 64

View Full Product Details