The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001211.5 | NP_001202.4 | 10 - 11 | 1608 | 141 |
GenBank mRNA | AB208782.1 | - | 10 - 11 | 1521 | 141 |
AF035933.1 | - | 10 - 11 | 1517 | 141 | |
AF046079.2 | - | 10 - 11 | 1438 | 141 | |
AF046918.1 | - | 10 - 11 | 1526 | 141 | |
AF053306.1 | - | 10 - 11 | 1580 | 141 | |
AF068760.1 | - | 10 - 11 | 1502 | 141 | |
AF107297.1 | - | 10 - 11 | 1533 | 141 | |
AK296795.1 | - | 10 - 11 | 1412 | 141 | |
AK296984.1 | - | 10 - 11 | 1592 | 141 | |
AK312709.1 | - | 10 - 11 | 1576 | 141 | |
BC018739.1 | - | 10 - 11 | 1530 | 141 |