The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001211.5 | NP_001202.4 | 17 - 18 | 2490 | 118 |
GenBank mRNA | AB208782.1 | - | 17 - 18 | 2403 | 118 |
AF035933.1 | - | 17 - 18 | 2399 | 118 | |
AF046079.2 | - | 17 - 18 | 2320 | 118 | |
AF046918.1 | - | 17 - 18 | 2408 | 118 | |
AF068760.1 | - | 17 - 18 | 2384 | 118 | |
AF107297.1 | - | 17 - 18 | 2415 | 118 | |
AK296795.1 | - | 17 - 18 | 2105 | 118 | |
AK296984.1 | - | 17 - 18 | 2474 | 118 | |
AK312709.1 | - | 17 - 18 | 2458 | 118 | |
BC018739.1 | - | 17 - 18 | 2412 | 118 |