The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173513.1 | NP_001166984.1 | 13 - 14 | 1752 | 95 |
NM_052883.1 | NP_443115.1 | 14 - 15 | 1860 | 95 | |
GenBank mRNA | AF133519.1 | - | 14 - 15 | 1528 | 95 |
AF171055.1 | - | 14 - 15 | 1534 | 95 | |
BC030028.2 | - | 14 - 15 | 1838 | 95 | |
BC050032.1 | - | 14 - 15 | 1847 | 95 |