The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004588.4 | NP_004579.1 | 1 - 2 | 266 | 92 |
GenBank mRNA | AF049498.1 | - | 1 - 2 | 247 | 92 |
AF107028.1 | - | 1 - 2 | 286 | 92 | |
AK308834.1 | - | 1 - 2 | 247 | 92 | |
AK313891.1 | - | 1 - 2 | 252 | 92 | |
AY358945.1 | - | 1 - 2 | 220 | 92 | |
BC036793.1 | - | 1 - 2 | 243 | 92 | |
BT019487.1 | - | 1 - 2 | 75 | 92 | |
DA290917.1 | - | 1 - 2 | 266 | 92 | |
U87555.1 | - | 1 - 2 | 188 | 92 |