Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNG
Entrez Gene ID
55787
Gene Name
taxilin gamma
Gene Aliases
CXorf15, ELRG, FIAT, LSR5, TXLNGX
Chromosome Location
Chr.X: 16786432 - 16844519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168683.1 NP_001162154.1 3 - 4 524 116
NM_018360.2 NP_060830.2 5 - 6 920 116
XM_017029631.1 XP_016885120.1 2 - 3 894 116
GenBank mRNA AY739713.1 - 3 - 4 474 116
BC101572.1 - 5 - 6 887 116
BC101576.1 - 5 - 6 887 116

View Full Product Details