Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYTL4
Entrez Gene ID
94121
Gene Name
synaptotagmin like 4
Gene Aliases
SLP4
Chromosome Location
Chr.X: 100671964 - 100732148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129896.2 NP_001123368.1 13 - 14 1600 110
NM_001174068.1 NP_001167539.1 12 - 13 1479 110
NM_080737.2 NP_542775.2 14 - 15 1648 110
XM_005262229.2 XP_005262286.1 12 - 13 1480 110
XM_011531068.2 XP_011529370.1 14 - 15 1677 110
XM_017029966.1 XP_016885455.1 14 - 15 1677 110
XM_017029967.1 XP_016885456.1 13 - 14 1569 110
XM_017029968.1 XP_016885457.1 12 - 13 1480 110
XM_017029969.1 XP_016885458.1 12 - 13 1454 110
XM_017029970.1 XP_016885459.1 12 - 13 1402 110
XM_017029971.1 XP_016885460.1 13 - 14 1580 110
GenBank mRNA AF302493.1 - 11 - 12 1368 110
AK292588.1 - 13 - 14 1569 110
AK313086.1 - 12 - 13 1454 110
BC014913.1 - 14 - 15 1624 110
BX537410.1 - 13 - 14 1567 110

View Full Product Details