Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL4
Entrez Gene ID
94121
Gene Name
synaptotagmin like 4
Gene Aliases
SLP4
Chromosome Location
Chr.X: 100671964 - 100732148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129896.2 NP_001123368.1 15 - 16 1869 98
NM_001174068.1 NP_001167539.1 14 - 15 1748 98
NM_080737.2 NP_542775.2 16 - 17 1917 98
XM_005262229.2 XP_005262286.1 14 - 15 1749 98
XM_011531068.2 XP_011529370.1 16 - 17 1946 98
XM_017029966.1 XP_016885455.1 16 - 17 1946 98
XM_017029967.1 XP_016885456.1 15 - 16 1838 98
XM_017029968.1 XP_016885457.1 14 - 15 1749 98
XM_017029969.1 XP_016885458.1 14 - 15 1723 98
XM_017029970.1 XP_016885459.1 14 - 15 1671 98
XM_017029971.1 XP_016885460.1 15 - 16 1849 98
GenBank mRNA AF302493.1 - 13 - 14 1637 98
AK292588.1 - 15 - 16 1838 98
AK313086.1 - 14 - 15 1723 98
BC014913.1 - 16 - 17 1893 98
BX537410.1 - 15 - 16 1836 98

View Full Product Details