Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP1R12C
Entrez Gene ID
54776
Gene Name
protein phosphatase 1 regulatory subunit 12C
Gene Aliases
LENG3, MBS85, p84, p85
Chromosome Location
Chr.19: 55090913 - 55117600 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271618.1 NP_001258547.1 14 - 15 1818 87
NM_017607.3 NP_060077.1 14 - 15 1821 87
XM_005259013.3 XP_005259070.1 14 - 15 1812 87
XM_011527045.2 XP_011525347.1 14 - 15 1810 87
GenBank mRNA AB209452.1 - 14 - 15 1631 87
AF312028.1 - 14 - 15 1782 87
AK027086.1 - 14 - 15 1456 87
AK297187.1 - 11 - 12 1104 87
AK297426.1 - 14 - 15 1670 87
AL137618.1 - 9 - 10 855 87
BC008030.1 - 5 - 6 508 87
BC010628.1 - 13 - 14 1599 87

View Full Product Details