The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039577.3 | NP_001034666.1 | 3 - 4 | 510 | 104 |
NM_001320153.1 | NP_001307082.1 | 3 - 4 | 510 | 104 | |
NM_001320155.1 | NP_001307084.1 | 3 - 4 | 510 | 104 | |
NM_030937.4 | NP_112199.2 | 3 - 4 | 510 | 104 | |
NR_135154.1 | 3 - 4 | 510 | 104 | ||
XM_011542219.2 | XP_011540521.1 | 1 - 2 | 101 | 104 | |
XM_017002420.1 | XP_016857909.1 | 3 - 4 | 508 | 104 | |
XM_017002421.1 | XP_016857910.1 | 3 - 4 | 494 | 104 | |
XM_017002422.1 | XP_016857911.1 | 3 - 4 | 507 | 104 | |
GenBank mRNA | AF251294.1 | - | 1 - 2 | 146 | 104 |
AK056120.1 | - | 3 - 4 | 483 | 104 | |
AK074112.1 | - | 3 - 4 | 481 | 104 | |
AK225758.1 | - | 3 - 4 | 505 | 104 | |
AL834441.1 | - | 3 - 4 | 441 | 104 | |
AY037150.1 | - | 3 - 4 | 484 | 104 | |
AY116620.1 | - | 3 - 4 | 499 | 104 | |
BC016333.1 | - | 3 - 4 | 490 | 104 |