The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320153.1 | NP_001307082.1 | 7 - 8 | 1512 | 85 |
NM_001320155.1 | NP_001307084.1 | 7 - 8 | 802 | 85 | |
NM_030937.4 | NP_112199.2 | 6 - 7 | 788 | 85 | |
NR_135154.1 | 6 - 7 | 2726 | 85 | ||
XM_011542216.2 | XP_011540518.1 | 6 - 7 | 863 | 85 | |
XM_011542219.2 | XP_011540521.1 | 6 - 7 | 672 | 85 | |
XM_011542221.2 | XP_011540523.1 | 5 - 6 | 503 | 85 | |
XM_017002420.1 | XP_016857909.1 | 7 - 8 | 2293 | 85 | |
XM_017002421.1 | XP_016857910.1 | 8 - 9 | 1176 | 85 | |
XM_017002422.1 | XP_016857911.1 | 7 - 8 | 910 | 85 | |
GenBank mRNA | AF251294.1 | - | 5 - 6 | 1241 | 85 |
AK000685.1 | - | 1 - 2 | 328 | 85 | |
AK026796.1 | - | 1 - 2 | 987 | 85 | |
AK027770.1 | - | 1 - 2 | 1757 | 85 | |
AK056120.1 | - | 7 - 8 | 1484 | 85 | |
AK057133.1 | - | 5 - 6 | 503 | 85 | |
AK074112.1 | - | 6 - 7 | 2697 | 85 | |
AK225758.1 | - | 6 - 7 | 783 | 85 | |
AK292268.1 | - | 1 - 2 | 311 | 85 | |
AK293451.1 | - | 3 - 4 | 266 | 85 | |
AY037150.1 | - | 6 - 7 | 762 | 85 | |
BC071622.1 | - | 1 - 2 | 175 | 85 |