The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025194.2 | NP_079470.1 | 3 - 4 | 1503 | 85 |
XM_017027324.1 | XP_016882813.1 | 4 - 5 | 687 | 85 | |
GenBank mRNA | AJ290975.1 | - | 3 - 4 | 1470 | 85 |
AK024596.1 | - | 3 - 4 | 538 | 85 | |
BC060788.1 | - | 3 - 4 | 1503 | 85 | |
D38169.1 | - | 3 - 4 | 1234 | 85 |