The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286526.1 | NP_001273455.1 | 3 - 4 | 637 | 73 |
NM_004765.3 | NP_004756.2 | 3 - 4 | 637 | 73 | |
XM_011545980.2 | XP_011544282.1 | 3 - 4 | 885 | 73 | |
XM_017023885.1 | XP_016879374.1 | 3 - 4 | 878 | 73 | |
XM_017023886.1 | XP_016879375.1 | 3 - 4 | 883 | 73 | |
XM_017023887.1 | XP_016879376.1 | 3 - 4 | 883 | 73 | |
XM_017023888.1 | XP_016879377.1 | 3 - 4 | 883 | 73 | |
GenBank mRNA | AJ223980.1 | - | 3 - 4 | 279 | 73 |
BC019071.1 | - | 3 - 4 | 413 | 73 | |
BC058863.1 | - | 3 - 4 | 637 | 73 |