The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077238.1 | NP_001070706.1 | 12 - 13 | 1360 | 74 |
NM_152988.2 | NP_694533.1 | 12 - 13 | 1360 | 74 | |
XM_011528138.2 | XP_011526440.1 | 12 - 13 | 1416 | 74 | |
XM_011528139.1 | XP_011526441.1 | 11 - 12 | 1218 | 74 | |
XM_011528140.1 | XP_011526442.1 | 8 - 9 | 771 | 74 | |
XM_017027017.1 | XP_016882506.1 | 13 - 14 | 1554 | 74 | |
XM_017027018.1 | XP_016882507.1 | 11 - 12 | 1215 | 74 | |
XM_017027019.1 | XP_016882508.1 | 13 - 14 | 1547 | 74 | |
GenBank mRNA | AB040965.1 | - | 12 - 13 | 1309 | 74 |
AJ345027.1 | - | 12 - 13 | 1313 | 74 | |
AJ420897.1 | - | 12 - 13 | 1309 | 74 | |
AY169315.1 | - | 12 - 13 | 1308 | 74 | |
BC028391.2 | - | 12 - 13 | 1297 | 74 | |
BC093046.1 | - | 12 - 13 | 1360 | 74 |