The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152988.2 | NP_694533.1 | 14 - 15 | 1595 | 110 |
XM_011528138.2 | XP_011526440.1 | 14 - 15 | 1651 | 110 | |
XM_011528139.1 | XP_011526441.1 | 13 - 14 | 1453 | 110 | |
XM_011528140.1 | XP_011526442.1 | 10 - 11 | 1006 | 110 | |
XM_017027017.1 | XP_016882506.1 | 15 - 16 | 1789 | 110 | |
XM_017027018.1 | XP_016882507.1 | 13 - 14 | 1450 | 110 | |
XM_017027019.1 | XP_016882508.1 | 15 - 16 | 1782 | 110 | |
GenBank mRNA | AB040965.1 | - | 14 - 15 | 1544 | 110 |
AJ345027.1 | - | 14 - 15 | 1548 | 110 | |
AJ420897.1 | - | 14 - 15 | 1544 | 110 | |
AY169315.1 | - | 14 - 15 | 1543 | 110 | |
BC093046.1 | - | 14 - 15 | 1595 | 110 |