The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002031.2 | NP_001002031.1 | 2 - 3 | 626 | 63 |
NM_005176.5 | NP_005167.2 | 2 - 3 | 346 | 63 | |
XM_017019460.1 | XP_016874949.1 | 3 - 4 | 573 | 63 | |
XM_017019461.1 | XP_016874950.1 | 3 - 4 | 357 | 63 | |
XM_017019462.1 | XP_016874951.1 | 2 - 3 | 2126 | 63 | |
GenBank mRNA | AK075351.1 | - | 1 - 2 | 1450 | 63 |
AK130971.1 | - | 2 - 3 | 103 | 63 | |
AK311357.1 | - | 3 - 4 | 1425 | 63 | |
BC013839.2 | - | 1 - 2 | 75 | 63 | |
BC020826.2 | - | 1 - 2 | 75 | 63 | |
BG217012.1 | - | 2 - 3 | 230 | 63 | |
BX348847.1 | - | 2 - 3 | 586 | 63 | |
D13119.1 | - | 1 - 2 | 72 | 63 |