The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242857.2 | NP_001229786.1 | 11 - 12 | 1019 | 93 |
NM_001242858.2 | NP_001229787.1 | 8 - 9 | 962 | 93 | |
NM_001242859.2 | NP_001229788.1 | 10 - 11 | 1010 | 93 | |
NM_001242860.2 | NP_001229789.1 | 11 - 12 | 1154 | 93 | |
NM_001242861.2 | NP_001229790.1 | 9 - 10 | 1046 | 93 | |
NM_001242862.2 | NP_001229791.1 | 7 - 8 | 758 | 93 | |
NM_002731.3 | NP_002722.1 | 8 - 9 | 1120 | 93 | |
NM_182948.3 | NP_891993.1 | 8 - 9 | 1024 | 93 | |
XM_005271015.3 | XP_005271072.1 | 10 - 11 | 971 | 93 | |
XM_005271016.1 | XP_005271073.1 | 10 - 11 | 1096 | 93 | |
XM_005271017.3 | XP_005271074.1 | 9 - 10 | 961 | 93 | |
XM_005271018.4 | XP_005271075.1 | 9 - 10 | 1281 | 93 | |
XM_005271019.1 | XP_005271076.1 | 8 - 9 | 1037 | 93 | |
XM_006710758.1 | XP_006710821.1 | 10 - 11 | 1110 | 93 | |
GenBank mRNA | AB209189.1 | - | 11 - 12 | 1090 | 93 |
AB451239.1 | - | 8 - 9 | 769 | 93 | |
AB451364.1 | - | 8 - 9 | 769 | 93 | |
AK091420.1 | - | 9 - 10 | 720 | 93 | |
AK296482.1 | - | 11 - 12 | 821 | 93 | |
AK304375.1 | - | 8 - 9 | 764 | 93 | |
AK314550.1 | - | 8 - 9 | 1016 | 93 | |
AK316115.1 | - | 7 - 8 | 731 | 93 | |
BC035058.1 | - | 8 - 9 | 93 | ||
BX537705.1 | - | 8 - 9 | 1001 | 93 | |
BX641026.1 | - | 8 - 9 | 1000 | 93 | |
CR936631.1 | - | 10 - 11 | 813 | 93 | |
M34181.1 | - | 8 - 9 | 816 | 93 |