Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCT2
Entrez Gene ID
10576
Gene Name
chaperonin containing TCP1 subunit 2
Gene Aliases
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta
Chromosome Location
Chr.12: 69585428 - 69601577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198842.1 NP_001185771.1 14 - 15 1759 129
NM_006431.2 NP_006422.1 14 - 15 1528 129
GenBank mRNA AB451223.1 - 13 - 14 1434 129
AB451346.1 - 13 - 14 1434 129
AF026166.1 - 14 - 15 1491 129
AF026293.1 - 14 - 15 1467 129
AK290767.1 - 14 - 15 1523 129
AK294307.1 - 14 - 15 1773 129
AK297685.1 - 12 - 13 1318 129
AK302157.1 - 14 - 15 1568 129
AK316397.1 - 14 - 15 1698 129
AK316408.1 - 14 - 15 1759 129
BC113514.1 - 14 - 15 1506 129
BC113516.1 - 14 - 15 1506 129
BT019966.1 - 13 - 14 1434 129
FJ224351.1 - 14 - 15 1467 129

View Full Product Details