The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 15 - 16 | 2211 | 87 |
GenBank mRNA | AF021351.1 | - | 16 - 17 | 2078 | 87 |
AK026023.1 | - | 6 - 7 | 785 | 87 | |
AK300329.1 | - | 15 - 16 | 2191 | 87 | |
AY091460.1 | - | 8 - 9 | 986 | 87 | |
BC041089.1 | - | 15 - 16 | 2172 | 87 |