Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3A
Entrez Gene ID
11128
Gene Name
RNA polymerase III subunit A
Gene Aliases
ADDH, HLD7, RPC1, RPC155, hRPC155
Chromosome Location
Chr.10: 77975149 - 78029540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007055.3 NP_008986.2 18 - 19 2613 88
GenBank mRNA AF021351.1 - 19 - 20 2480 88
AK026023.1 - 9 - 10 1187 88
AK300329.1 - 18 - 19 2593 88
AY091461.1 - 3 - 4 287 88
BC041089.1 - 18 - 19 2574 88

View Full Product Details