Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR3A
Entrez Gene ID
11128
Gene Name
RNA polymerase III subunit A
Gene Aliases
ADDH, HLD7, RPC1, RPC155, hRPC155
Chromosome Location
Chr.10: 77975149 - 78029540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007055.3 NP_008986.2 7 - 8 1181 104
GenBank mRNA AF021351.1 - 8 - 9 1048 104
AK300329.1 - 7 - 8 1161 104
AY091459.1 - 7 - 8 1048 104
BC041089.1 - 7 - 8 1142 104

View Full Product Details