Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D15
Entrez Gene ID
64786
Gene Name
TBC1 domain family member 15
Gene Aliases
RAB7-GAP
Chromosome Location
Chr.12: 71839707 - 71926849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146213.1 NP_001139685.2 5 - 6 626 161
NM_001146214.1 NP_001139686.1 6 - 7 718 161
NM_022771.4 NP_073608.4 5 - 6 626 161
XM_006719564.2 XP_006719627.1 5 - 6 608 161
XM_006719565.3 XP_006719628.1 5 - 6 546 161
XM_006719566.2 XP_006719629.1 6 - 7 757 161
XM_011538679.1 XP_011536981.1 7 - 8 683 161
XM_011538680.1 XP_011536982.1 5 - 6 641 161
XM_011538681.2 XP_011536983.1 5 - 6 590 161
XM_017019824.1 XP_016875313.1 6 - 7 643 161
XM_017019825.1 XP_016875314.1 6 - 7 713 161
XM_017019826.1 XP_016875315.1 6 - 7 643 161
XM_017019827.1 XP_016875316.1 2 - 3 266 161
XM_017019828.1 XP_016875317.1 4 - 5 435 161
GenBank mRNA AB449901.1 - 5 - 6 551 161
AK022147.1 - 5 - 6 574 161
AK225221.1 - 5 - 6 555 161
AK225747.1 - 5 - 6 572 161
AK297626.1 - 6 - 7 666 161
AK307922.1 - 6 - 7 643 161
BC028352.2 - 5 - 6 574 161

View Full Product Details