The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005055.4 | NP_005046.2 | 5 - 6 | 1130 | 78 |
GenBank mRNA | AF449218.1 | - | 5 - 6 | 1130 | 78 |
KU178301.1 | - | 5 - 6 | 916 | 78 | |
KU178302.1 | - | 5 - 6 | 916 | 78 | |
Z33905.1 | - | 5 - 6 | 1123 | 78 |