The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144699.3 | NP_653300.2 | 11 - 12 | 2160 | 130 |
XM_011509582.1 | XP_011507884.1 | 10 - 11 | 1627 | 130 | |
GenBank mRNA | AF506797.1 | - | 11 - 12 | 2152 | 130 |
AL833335.1 | - | 11 - 12 | 2133 | 130 | |
BC094801.1 | - | 11 - 12 | 2111 | 130 | |
BX647234.1 | - | 11 - 12 | 2134 | 130 |