The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001734.1 | NP_001001734.1 | 2 - 3 | 385 | 68 |
NM_144699.3 | NP_653300.2 | 19 - 20 | 3346 | 68 | |
XM_011509582.1 | XP_011507884.1 | 18 - 19 | 2813 | 68 | |
GenBank mRNA | AF506797.1 | - | 19 - 20 | 3338 | 68 |
AK098076.1 | - | 9 - 10 | 1223 | 68 | |
AL833335.1 | - | 19 - 20 | 3319 | 68 | |
BC028297.1 | - | 2 - 3 | 385 | 68 | |
BC094801.1 | - | 19 - 20 | 3297 | 68 | |
BX647234.1 | - | 19 - 20 | 3320 | 68 |