The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099660.1 | NP_001093130.1 | 1 - 2 | 495 | 90 |
GenBank mRNA | AK001991.1 | - | 1 - 2 | 407 | 90 |