The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017373.3 | NP_001017373.2 | 5 - 6 | 707 | 114 |
NM_001258275.2 | NP_001245204.1 | 7 - 8 | 988 | 114 | |
NM_001277185.1 | NP_001264114.1 | 4 - 5 | 489 | 114 | |
XM_017010306.1 | XP_016865795.1 | 3 - 4 | 747 | 114 | |
XM_017010308.1 | XP_016865797.1 | 3 - 4 | 400 | 114 | |
XM_017010309.1 | XP_016865798.1 | 3 - 4 | 400 | 114 | |
XM_017010310.1 | XP_016865799.1 | 3 - 4 | 400 | 114 | |
GenBank mRNA | AK091351.1 | - | 7 - 8 | 990 | 114 |
AK091882.1 | - | 5 - 6 | 707 | 114 | |
AK302228.1 | - | 4 - 5 | 491 | 114 | |
AK310075.1 | - | 3 - 4 | 743 | 114 | |
BC029851.1 | - | 3 - 4 | 396 | 114 | |
BC119757.1 | - | 3 - 4 | 396 | 114 | |
BC119758.1 | - | 3 - 4 | 396 | 114 | |
BC127653.1 | - | 3 - 4 | 394 | 114 | |
BC127654.1 | - | 3 - 4 | 394 | 114 |