Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
Chromosome Location
Chr.12: 8102932 - 8138607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016184.3 NP_057268.1 5 - 6 810 68
NM_194447.2 NP_919429.2 4 - 5 693 68
NM_194448.2 NP_919430.1 3 - 4 594 68
NM_194450.2 NP_919432.1 4 - 5 711 68
XM_011520684.2 XP_011518986.1 4 - 5 2071 68
XM_017019382.1 XP_016874871.1 6 - 7 731 68
GenBank mRNA AA418441.1 - 1 - 2 46 68
AF067800.1 - 5 - 6 831 68
AF109146.1 - 5 - 6 657 68
AF200738.1 - 4 - 5 464 68
AF328684.1 - 5 - 6 802 68
AI400631.1 - 1 - 2 68
AJ133532.1 - 5 - 6 804 68
BC074822.2 - 5 - 6 585 68
BC074874.2 - 5 - 6 585 68
BC117439.1 - 5 - 6 705 68
BC117441.1 - 5 - 6 705 68
EU196761.1 - 3 - 4 279 68

View Full Product Details