The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000553.4 | NP_000544.2 | 13 - 14 | 2436 | 98 |
XM_011544639.2 | XP_011542941.1 | 12 - 13 | 1837 | 98 | |
GenBank mRNA | AF091214.1 | - | 13 - 14 | 1879 | 98 |
AY818673.1 | - | 12 - 13 | 1648 | 98 |