Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
Chromosome Location
Chr.8: 31033262 - 31173761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000553.4 NP_000544.2 33 - 34 4765 123
XM_011544639.2 XP_011542941.1 32 - 33 4166 123
XM_011544640.1 XP_011542942.1 21 - 22 2478 123
GenBank mRNA AB209652.1 - 19 - 20 2208 123
AF091214.1 - 33 - 34 4208 123
AY818673.1 - 32 - 33 3977 123

View Full Product Details