The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033358.1 | 5 - 6 | 1178 | 110 | |
NR_033360.1 | 4 - 5 | 1064 | 110 | ||
NR_046467.1 | 5 - 6 | 1216 | 110 | ||
NR_046472.1 | 4 - 5 | 1102 | 110 | ||
GenBank mRNA | GQ183494.1 | - | 4 - 5 | 1056 | 110 |
GQ183498.1 | - | 5 - 6 | 1208 | 110 | |
GQ183499.1 | - | 5 - 6 | 1171 | 110 | |
GQ183503.1 | - | 4 - 5 | 1094 | 110 |