The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077399.2 | NP_001070867.1 | 2 - 3 | 380 | 73 |
GenBank mRNA | AF318057.1 | - | 2 - 3 | 380 | 73 |
AF417001.1 | - | 2 - 3 | 380 | 73 | |
AL080092.1 | - | 2 - 3 | 262 | 73 | |
AY039043.1 | - | - | 268 | 73 | |
BC021118.1 | - | 2 - 3 | 267 | 73 | |
BQ028777.1 | - | 1 - 2 | 73 |