The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144629.2 | NP_653230.2 | 1 - 2 | 546 | 168 |
XM_011510595.2 | XP_011508897.1 | 1 - 2 | 736 | 168 | |
XM_011510596.2 | XP_011508898.1 | 1 - 2 | 736 | 168 | |
XM_011510597.2 | XP_011508899.1 | 1 - 2 | 736 | 168 | |
XM_011510598.2 | XP_011508900.1 | 1 - 2 | 736 | 168 | |
XM_017003332.1 | XP_016858821.1 | 1 - 2 | 736 | 168 | |
GenBank mRNA | AK055136.1 | - | 1 - 2 | 546 | 168 |
BC093966.1 | - | 1 - 2 | 193 | 168 | |
BC111069.1 | - | 1 - 2 | 678 | 168 | |
BC113361.1 | - | 1 - 2 | 193 | 168 |