Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LARS
Entrez Gene ID
51520
Gene Name
leucyl-tRNA synthetase
Gene Aliases
HSPC192, ILFS1, LARS1, LEURS, LEUS, LFIS, LRS, PIG44, RNTLS, hr025Cl
Chromosome Location
Chr.5: 146113026 - 146182787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317964.1 NP_001304893.1 10 - 11 1308 76
NM_001317965.1 NP_001304894.1 10 - 11 1327 76
NM_016460.3 NP_057544.2 10 - 11 1365 76
NM_020117.10 NP_064502.9 11 - 12 1446 76
XM_011537656.2 XP_011535958.1 11 - 12 1312 76
GenBank mRNA AB037773.1 - 11 - 12 1319 76
AK023777.1 - 4 - 5 430 76
AK025441.1 - 11 - 12 1282 76
AK225372.1 - 10 - 11 1144 76
AK293750.1 - 10 - 11 1195 76
AK295874.1 - 10 - 11 1163 76
AK304134.1 - 10 - 11 1124 76
AK315178.1 - 11 - 12 1295 76
BC150213.1 - 11 - 12 1297 76
BC151214.1 - 11 - 12 1247 76
BC152422.1 - 11 - 12 1319 76
D84223.1 - 11 - 12 1224 76

View Full Product Details