The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146337.2 | NP_001139809.1 | 2 - 3 | 274 | 82 |
NM_001323062.1 | NP_001309991.1 | 2 - 3 | 274 | 82 | |
NM_001323063.1 | NP_001309992.1 | 2 - 3 | 274 | 82 | |
NM_152406.3 | NP_689619.1 | 2 - 3 | 274 | 82 | |
XM_017009036.1 | XP_016864525.1 | 2 - 3 | 251 | 82 | |
GenBank mRNA | AK074185.1 | - | 2 - 3 | 217 | 82 |
AK095980.1 | - | 2 - 3 | 228 | 82 | |
BC040723.2 | - | 2 - 3 | 241 | 82 | |
BC125093.1 | - | 2 - 3 | 193 | 82 | |
BC125094.1 | - | 2 - 3 | 193 | 82 | |
DA792192.1 | - | 2 - 3 | 274 | 82 |