The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083617.1 | NP_001077086.1 | 2 - 3 | 503 | 104 |
NM_014781.4 | NP_055596.3 | 2 - 3 | 503 | 104 | |
XM_011517644.2 | XP_011515946.1 | 2 - 3 | 503 | 104 | |
XM_011517645.1 | XP_011515947.1 | 2 - 3 | 503 | 104 | |
XM_011517647.2 | XP_011515949.1 | 2 - 3 | 503 | 104 | |
XM_011517649.2 | XP_011515951.1 | 2 - 3 | 503 | 104 | |
XM_017014103.1 | XP_016869592.1 | 2 - 3 | 503 | 104 | |
XM_017014105.1 | XP_016869594.1 | 2 - 3 | 503 | 104 | |
XM_017014106.1 | XP_016869595.1 | 2 - 3 | 503 | 104 | |
XM_017014108.1 | XP_016869597.1 | 2 - 3 | 503 | 104 | |
XM_017014110.1 | XP_016869599.1 | 2 - 3 | 503 | 104 | |
XM_017014111.1 | XP_016869600.1 | 2 - 3 | 503 | 104 | |
XM_017014112.1 | XP_016869601.1 | 2 - 3 | 503 | 104 | |
GenBank mRNA | AB059622.1 | - | 2 - 3 | 469 | 104 |
AK291699.1 | - | 2 - 3 | 460 | 104 | |
D86958.1 | - | 2 - 3 | 460 | 104 |