The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014737.2 | NP_055552.1 | 4 - 5 | 327 | 92 |
NM_170774.1 | NP_739580.1 | 3 - 4 | 185 | 92 | |
XM_005260895.3 | XP_005260952.1 | 4 - 5 | 393 | 92 | |
XM_011529410.1 | XP_011527712.1 | 4 - 5 | 321 | 92 | |
XM_011529411.1 | XP_011527713.1 | 4 - 5 | 393 | 92 | |
XM_011529412.2 | XP_011527714.1 | 3 - 4 | 242 | 92 | |
XM_017028149.1 | XP_016883638.1 | 3 - 4 | 216 | 92 | |
XM_017028150.1 | XP_016883639.1 | 5 - 6 | 530 | 92 | |
XM_017028151.1 | XP_016883640.1 | 5 - 6 | 497 | 92 | |
XM_017028152.1 | XP_016883641.1 | 5 - 6 | 574 | 92 | |
XM_017028153.1 | XP_016883642.1 | 3 - 4 | 275 | 92 | |
GenBank mRNA | AK222650.1 | - | 4 - 5 | 290 | 92 |
AK291458.1 | - | 4 - 5 | 282 | 92 | |
AY154470.1 | - | 4 - 5 | 327 | 92 | |
AY154472.1 | - | 3 - 4 | 185 | 92 | |
BC117118.1 | - | 2 - 3 | 164 | 92 | |
BC117120.1 | - | 2 - 3 | 164 | 92 | |
CR627436.1 | - | 4 - 5 | 347 | 92 | |
D79990.1 | - | 4 - 5 | 327 | 92 | |
DA333367.1 | - | 4 - 5 | 290 | 92 |