The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167911.1 | NP_001161383.1 | 6 - 7 | 1135 | 68 |
NM_001167912.1 | NP_001161384.1 | 6 - 7 | 1135 | 68 | |
NM_024621.2 | NP_078897.2 | 6 - 7 | 1326 | 68 | |
XM_011513134.2 | XP_011511436.1 | 5 - 6 | 1069 | 68 | |
XM_011513135.1 | XP_011511437.1 | 6 - 7 | 965 | 68 | |
GenBank mRNA | AB051479.1 | - | 6 - 7 | 1135 | 68 |
AK027625.1 | - | 6 - 7 | 1224 | 68 | |
AL713656.1 | - | 5 - 6 | 923 | 68 | |
BC101660.1 | - | 5 - 6 | 962 | 68 | |
BC111017.1 | - | 6 - 7 | 1127 | 68 | |
BC113555.1 | - | 5 - 6 | 962 | 68 |