Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSRP2
Entrez Gene ID
1466
Gene Name
cysteine and glycine rich protein 2
Gene Aliases
CRP2, LMO5, SmLIM
Chromosome Location
Chr.12: 76858715 - 76879040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300965.1 NP_001287894.1 4 - 5 495 102
NM_001321.2 NP_001312.1 4 - 5 520 102
GenBank mRNA AB209321.1 - 1 - 2 4672 102
BC000992.2 - 4 - 5 468 102
BT019913.1 - 3 - 4 416 102
CN408330.1 - 4 - 5 466 102
U46006.1 - 3 - 4 416 102
U57646.1 - 4 - 5 472 102

View Full Product Details