Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMBP
Entrez Gene ID
259
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI
Chromosome Location
Chr.9: 114060127 - 114078472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001633.3 NP_001624.1 7 - 8 949 66
GenBank mRNA AK290837.1 - 7 - 8 777 66
AK303795.1 - 7 - 8 675 66
AY544123.1 - 7 - 8 746 66
BC041593.1 - 7 - 8 912 66
X04225.1 - 7 - 8 758 66
X04494.1 - 7 - 8 731 66

View Full Product Details