Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMBP
Entrez Gene ID
259
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI
Chromosome Location
Chr.9: 114060127 - 114078472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001633.3 NP_001624.1 8 - 9 1119 75
GenBank mRNA AK290837.1 - 8 - 9 947 75
AY544123.1 - 8 - 9 916 75
BC041593.1 - 8 - 9 1082 75
X04225.1 - 8 - 9 928 75
X04494.1 - 8 - 9 901 75

View Full Product Details