The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301855.1 | NP_001288784.1 | 2 - 3 | 473 | 121 |
NM_013260.7 | NP_037392.1 | 2 - 3 | 473 | 121 | |
NR_126036.1 | 2 - 3 | 473 | 121 | ||
NR_126037.1 | 2 - 3 | 473 | 121 | ||
XM_011524693.2 | XP_011522995.1 | 2 - 3 | 261 | 121 | |
XM_011524695.2 | XP_011522997.1 | 2 - 3 | 252 | 121 | |
XM_017024546.1 | XP_016880035.1 | 1 - 2 | 110 | 121 | |
XM_017024548.1 | XP_016880037.1 | 2 - 3 | 252 | 121 | |
XM_017024549.1 | XP_016880038.1 | 2 - 3 | 252 | 121 | |
XM_017024550.1 | XP_016880039.1 | 2 - 3 | 219 | 121 | |
GenBank mRNA | AF087869.1 | - | 2 - 3 | 216 | 121 |
AF119664.1 | - | 2 - 3 | 231 | 121 | |
AF450482.1 | - | 2 - 3 | 473 | 121 | |
AK002202.1 | - | 2 - 3 | 223 | 121 | |
AK023157.1 | - | 2 - 3 | 200 | 121 | |
AK298060.1 | - | 2 - 3 | 233 | 121 | |
AK310177.1 | - | 2 - 3 | 223 | 121 | |
AY082382.1 | - | 2 - 3 | 231 | 121 | |
BC007592.1 | - | 2 - 3 | 227 | 121 | |
BC011724.2 | - | 2 - 3 | 218 | 121 | |
BC013409.2 | - | 2 - 3 | 203 | 121 | |
BC030233.1 | - | 2 - 3 | 233 | 121 | |
BC063793.1 | - | 2 - 3 | 225 | 121 | |
BM807177.1 | - | 2 - 3 | 163 | 121 |