The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301839.1 | NP_001288768.1 | 5 - 6 | 696 | 128 |
NM_001301855.1 | NP_001288784.1 | 7 - 8 | 846 | 128 | |
NM_013260.7 | NP_037392.1 | 6 - 7 | 744 | 128 | |
XM_011524693.2 | XP_011522995.1 | 6 - 7 | 532 | 128 | |
XM_011524694.2 | XP_011522996.1 | 5 - 6 | 478 | 128 | |
XM_011524695.2 | XP_011522997.1 | 8 - 9 | 767 | 128 | |
XM_017024545.1 | XP_016880034.1 | 5 - 6 | 439 | 128 | |
XM_017024546.1 | XP_016880035.1 | 7 - 8 | 625 | 128 | |
XM_017024547.1 | XP_016880036.1 | 5 - 6 | 436 | 128 | |
XM_017024548.1 | XP_016880037.1 | 8 - 9 | 763 | 128 | |
XM_017024549.1 | XP_016880038.1 | 8 - 9 | 763 | 128 | |
XM_017024550.1 | XP_016880039.1 | 6 - 7 | 490 | 128 | |
GenBank mRNA | AF087869.1 | - | 6 - 7 | 487 | 128 |
AF119664.1 | - | 6 - 7 | 502 | 128 | |
AF450482.1 | - | 6 - 7 | 744 | 128 | |
AK002202.1 | - | 6 - 7 | 494 | 128 | |
AK023157.1 | - | 6 - 7 | 471 | 128 | |
AK095265.1 | - | 5 - 6 | 466 | 128 | |
AY082382.1 | - | 6 - 7 | 502 | 128 | |
BC007592.1 | - | 6 - 7 | 498 | 128 | |
BC013409.2 | - | 6 - 7 | 474 | 128 | |
BC030233.1 | - | 6 - 7 | 504 | 128 | |
BC063793.1 | - | 6 - 7 | 496 | 128 | |
BM807177.1 | - | 7 - 8 | 536 | 128 | |
BX647066.1 | - | 3 - 4 | 2093 | 128 |